Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-fluorotoluene, 98+%

Catalog No. AAA1279214
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1279214 25 g
AAA1279206 5 g
AAA1279222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1279214 Supplier Thermo Scientific Chemicals Supplier No. A1279214
Add to Cart

CAS: 1422-53-3 | C7H6BrF | 189.027 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1422-53-3
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00040828
InChI Key SFGFOJPGCOYQJK-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorotoluene, 4-fluoro-2-bromotoluene, 2-bromo-4-fluoro toluene, benzene, 2-bromo-4-fluoro-1-methyl, 2-bromo-4-fluoro-1-methyl-benzene, 2-brom-4-fluor-1-methylbenzol, pubchem1602, 2-bromo4-fluorotoluene, acmc-209cny, 2-bromo-4-fluoro-toluene
PubChem CID 74016
IUPAC Name 2-bromo-4-fluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)Br

Specifications

Density 1.526
Boiling Point 170°C to 172°C
Flash Point 64°C (147°F)
Refractive Index 1.528
Quantity 25 g
Beilstein 1859038
Formula Weight 189.03
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-4-fluorotoluene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 215-830-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.