Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4'-hydroxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. B411525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B411525G 25 g
B41155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B411525G Supplier TCI America Supplier No. B411525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2491-38-5
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00072424
InChI Key LJYOFQHKEWTQRH-UHFFFAOYSA-N
Synonym 4-(Bromoacetyl)phenol, 4-Hydroxyphenacyl Bromide
PubChem CID 4964
IUPAC Name 2-bromo-1-(4-hydroxyphenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)O

Specifications

Melting Point 131°C
Quantity 25 g
UN Number 3261
Formula Weight 215.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4′-hydroxyacetophenone
RTECSNumber AM5982000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.