Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Bromo-4-methylphenol, 97%
Click to view available options
Quantity:
5 g
25 g
Description
It is applied as a compound responsible for iodine off-flavor in wines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as a compound responsible for iodine off-flavor in wines.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is applied as a compound responsible for iodine off-flavor in wines.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | 16°C |
Density | 1.547 |
Boiling Point | 213°C to 214°C |
Flash Point | >110°C (230°F) |
Odor | Phenol-like |
Refractive Index | 1.578 |
Quantity | 25 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 187.04 |
Percent Purity | 97% |
Show More |
Safety and Handling
Hazard Category | H302+H312-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H312-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 229-595-7 |
RTECSNumber | GO6868200 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction