Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-4'-methylpropiophenone, 98%

Click to view available options
Quantity:
1g
5g
25g
1451-82-7
C10H11BrO
227.101
MFCD11131402
OZLUPIIIHOOPNQ-UHFFFAOYSA-N
2-bromo-1-p-tolyl propan-1-one,2-bromo-4'-methylpropiophenone,2-bromo-1-4-methylphenyl propan-1-one,2-bromo-4-methylpropiophenone,2-bromo-1-4-methylphenyl-1-propanone,1-propanone, 2-bromo-1-4-methylphenyl,2-bromo-1-p-tolylpropan-1-one,ksc917e0p,2-bromo-1-p-tolyl-propan-1-one,4'-methyl-alpha-bromopropiophenone
2734063
2-bromo-1-(4-methylphenyl)propan-1-one
CC1=CC=C(C=C1)C(=O)C(C)Br
1451-82-7
C10H11BrO
227.101
MFCD11131402
OZLUPIIIHOOPNQ-UHFFFAOYSA-N
2-bromo-1-p-tolyl propan-1-one,2-bromo-4'-methylpropiophenone,2-bromo-1-4-methylphenyl propan-1-one,2-bromo-4-methylpropiophenone,2-bromo-1-4-methylphenyl-1-propanone,1-propanone, 2-bromo-1-4-methylphenyl,2-bromo-1-p-tolylpropan-1-one,ksc917e0p,2-bromo-1-p-tolyl-propan-1-one,4'-methyl-alpha-bromopropiophenone
2734063
2-bromo-1-(4-methylphenyl)propan-1-one
CC1=CC=C(C=C1)C(=O)C(C)Br
Applications
It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.

Solubility
Soluble in chloroform and methanol.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Quantity 25g
Beilstein 2207628
Formula Weight 227.1
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Bromo-4'-methylpropiophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning