Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-methylpyridine, 99%

Catalog No. AC350350100
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle

CAS: 4926-28-7 | C6H6BrN | 172.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4926-28-7
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00082590
InChI Key LSZMVESSGLHDJE-UHFFFAOYSA-N
Synonym 2-bromo-4-picoline, 2-bromo-4-methyl-pyridine, 4-methyl-2-bromopyridine, pyridine, 2-bromo-4-methyl, 2 bromo-4-methyl pyridine, 2-bromo-4-methyl pyridine, pubchem1184, acmc-209kfi, 2-bromo4-methylpyridine
PubChem CID 2734087
IUPAC Name 2-bromo-4-methylpyridine
SMILES CC1=CC=NC(Br)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.5450g/mL
Boiling Point 87°C (10.0 mmHg)
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5600 to 1.563
Quantity 10 g
Specific Gravity 1.545
Formula Weight 172.02
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-methylpyridine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.