Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-nitroanisole 98.0+%, TCI America™
SDP

Catalog No. B283025G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5197-28-4
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.03
MDL Number MFCD00041242
InChI Key JMUDXQVNBZCQRF-UHFFFAOYSA-N
Synonym 2-bromo-4-nitroanisole, 3-bromo-4-methoxynitrobenzene, anisole, 2-bromo-4-nitro, benzene, 2-bromo-1-methoxy-4-nitro, 2-bromo-1-methoxy-4-nitro-benzene, pubchem4133, 2-bromo4-nitroanisole, 4-nitro-2-bromanisol, acmc-1awkm, 2-bromo-4-nitro anisole
PubChem CID 78870
IUPAC Name 2-bromo-1-methoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1Br)[N+]([O-])=O

Specifications

Melting Point 104°C
Quantity 25 g
Formula Weight 232.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-nitroanisole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.