Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B360725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B360725G 25 g
B36075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B360725G Supplier TCI America Supplier No. B360725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16426-64-5
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
InChI Key CEXGTXNIIFSPSF-UHFFFAOYSA-N
PubChem CID 313534
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Br)C(=O)O

Specifications

Melting Point 169°C
Color Yellow
Quantity 25 g
Formula Weight 246.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.