Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4'-phenylacetophenone, 97%

Catalog No. p-7027502
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 135-73-9 | C14H11BrO | 275.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-73-9
Molecular Formula C14H11BrO
Molecular Weight (g/mol) 275.145
MDL Number MFCD00000202
InChI Key KGHGZRVXCKCJGX-UHFFFAOYSA-N
Synonym 2-bromo-4'-phenylacetophenone, 4-phenylphenacyl bromide, p-bromoacetylbiphenyl, p-phenylphenacyl bromide, bromomethyl p-biphenylyl ketone, ethanone, 1-1,1'-biphenyl-4-yl-2-bromo, 2-bromo-1-4-phenylphenyl ethan-1-one, acetophenone, 2-bromo-4'-phenyl, alpha-bromo-p-phenylacetophenone, omega-bromo-4-phenylacetophenone
PubChem CID 67282
IUPAC Name 2-bromo-1-(4-phenylphenyl)ethanone
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CBr

Specifications

Melting Point 123°C to 127°C
Quantity 5 g
UN Number UN3261
Beilstein 513838
Sensitivity Light sensitive
Formula Weight 275.15
Percent Purity 97%
Chemical Name or Material 2-Bromo-4'-phenylacetophenone
Hazard Category H314-H335-H500
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 205-217-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.