Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4'-phenylacetophenone 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
135-73-9
C14H11BrO
275.13
MFCD00000202
KGHGZRVXCKCJGX-UHFFFAOYSA-N
2-bromo-4'-phenylacetophenone,4-phenylphenacyl bromide,p-bromoacetylbiphenyl,p-phenylphenacyl bromide,bromomethyl p-biphenylyl ketone,ethanone, 1-1,1'-biphenyl-4-yl-2-bromo,2-bromo-1-4-phenylphenyl ethan-1-one,acetophenone, 2-bromo-4'-phenyl,alpha-bromo-p-phenylacetophenone,omega-bromo-4-phenylacetophenone
67282
2-bromo-1-(4-phenylphenyl)ethanone
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CBr
135-73-9
C14H11BrO
275.13
MFCD00000202
KGHGZRVXCKCJGX-UHFFFAOYSA-N
2-bromo-4'-phenylacetophenone,4-phenylphenacyl bromide,p-bromoacetylbiphenyl,p-phenylphenacyl bromide,bromomethyl p-biphenylyl ketone,ethanone, 1-1,1'-biphenyl-4-yl-2-bromo,2-bromo-1-4-phenylphenyl ethan-1-one,acetophenone, 2-bromo-4'-phenyl,alpha-bromo-p-phenylacetophenone,omega-bromo-4-phenylacetophenone
67282
2-bromo-1-(4-phenylphenyl)ethanone
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CBr
Packaging Glass bottle
Color Undesignated
Melting Point 123°C to 127°C
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5C6H4COCH2Br
Beilstein 07, III, 2137
Formula Weight 275.13
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Bromo-4'-phenylacetophenone, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 205-217-6

TSCA : TSCA