Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-decylthiophene 97.0+%, TCI America™
SDP

Catalog No. B54061G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 514188-72-8
Molecular Formula C14H23BrS
Molecular Weight (g/mol) 303.30
MDL Number MFCD30742895
InChI Key BLGUNQPKOOEOHU-UHFFFAOYSA-N
PubChem CID 58892816
IUPAC Name 2-bromo-5-decylthiophene
SMILES CCCCCCCCCCC1=CC=C(Br)S1

Specifications

Color Yellow
Boiling Point 135°C
Quantity 1 g
Formula Weight 303.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-decylthiophene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.