Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-dodecylthiophene 95.0+%, TCI America™
SDP

Catalog No. B54071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54071G 1 g
B54075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54071G Supplier TCI America Supplier No. B54071G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 153561-74-1
Molecular Formula C16H27BrS
Molecular Weight (g/mol) 331.36
MDL Number MFCD22572689
InChI Key MOSAVYMGWTYDNS-UHFFFAOYSA-N
PubChem CID 14973118
IUPAC Name 2-bromo-5-dodecylthiophene
SMILES CCCCCCCCCCCCC1=CC=C(Br)S1

Specifications

Color Yellow
Boiling Point 162°C
Quantity 1 g
Formula Weight 331.36
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-dodecylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.