Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-fluoroaniline, 98%

Catalog No. p-7027852
Click to view available options
Quantity:
5 g
25 g

CAS: 1003-99-2 | C6H5BrFN | 190.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-99-2
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD00070750
InChI Key FWTXFEKVHSFTDQ-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-5-fluoro, 2-bromo-5-fluorobenzenamine, 2-bromo-5-fluoro-aniline, 2-bromo-5-fluoro-phenylamine, 2-bromo-5-fluoro-benzenamine, 2-bromo-5-fluorophenylamine, pubchem1524, 5-fluoro-2-bromoaniline, 2-bromo-5-fluoro aniline, acmc-1c3a5
PubChem CID 2773317
IUPAC Name 2-bromo-5-fluoroaniline
SMILES C1=CC(=C(C=C1F)N)Br

Specifications

Melting Point 42°C to 46°C
Boiling Point 154°C (23 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2811
Formula Weight 190.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-fluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.