Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-fluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B272225G
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 394-28-5
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD00142874
InChI Key OQBMJMJZMDBQSM-UHFFFAOYSA-M
Synonym benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747
PubChem CID 2778181
IUPAC Name 2-bromo-5-fluorobenzoate
SMILES [O-]C(=O)C1=CC(F)=CC=C1Br

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 25 g
Formula Weight 219.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-fluorobenzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.