Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-fluorobenzotrifluoride 97.0+%, TCI America™
SDP

Catalog No. B280025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B280025G 25 g
B28005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B280025G Supplier TCI America Supplier No. B280025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40161-55-5
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243.00
MDL Number MFCD00040937
InChI Key AIDVAZGOACECLJ-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorobenzotrifluoride, 1-bromo-4-fluoro-2-trifluoromethyl benzene, 1-bromo-2-trifluoromethyl-4-fluorobenzene, 4-fluoro-2-trifluoromethyl bromobenzene, 2-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, benzene, 1-bromo-4-fluoro-2-trifluoromethyl, 2-bromo-5-fluoro benzotrifluoride, pubchem1637, pubchem1643
PubChem CID 520983
IUPAC Name 1-bromo-4-fluoro-2-(trifluoromethyl)benzene
SMILES FC1=CC(=C(Br)C=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 143°C
Quantity 25 g
Formula Weight 243.00
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-fluorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.