Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-fluorophenyl acetic acid, 96%, Thermo Scientific Chemicals

Catalog No. AAH6200503
Click to view available options
Quantity:
1 g
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 739336-26-6
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.036
MDL Number MFCD03094440
InChI Key BPNWLZNAKZSBHH-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorophenylacetic acid, 2-2-bromo-5-fluorophenyl acetic acid, 2-bromo-5-fluorophenyl acetic acid, 2-bromo-5-fluorophenylaceticacid, 2-bromo-5-fulorophenylacetic acid, benzeneacetic acid, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenyl-acetic acid, pubchem16327, 2-2-bromo-5-fluoro-phenyl acetic acid, acmc-209osx
PubChem CID 2773927
IUPAC Name 2-(2-bromo-5-fluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1F)CC(=O)O)Br

Specifications

Melting Point 113°C to 114°C
Quantity 1 g
Formula Weight 233.03
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-fluorophenyl acetic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.