Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-methoxybenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. B442325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B442325G 25 g
B44235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B442325G Supplier TCI America Supplier No. B442325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19614-12-1
Molecular Formula C8H8Br2O
Molecular Weight (g/mol) 279.959
MDL Number MFCD00078664
InChI Key MURVUTUZSUEIGI-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzyl bromide, 1-bromo-2-bromomethyl-4-methoxybenzene, 2-bromo-5-methoxybenzylbromide, 4-bromo-3-bromomethyl anisole, 2-bromo-5-methoxy-benzyl bromide, benzene, 1-bromo-2-bromomethyl-4-methoxy, 4-bromo-3-bromomethyl-1-methoxybenzene, pubchem18176, acmc-20a5x2, 4-bromo3-bromomethylanisole
PubChem CID 3525951
IUPAC Name 1-bromo-2-(bromomethyl)-4-methoxybenzene
SMILES COC1=CC(=C(C=C1)Br)CBr

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 279.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-methoxybenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.