Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-methylpyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
3510-66-5
C6H6BrN
172.02
MFCD00209553
YWNJQQNBJQUKME-UHFFFAOYSA-N
2-bromo-5-picoline,6-bromo-3-picoline,2-bromo-5-methyl-pyridine,pyridine, 2-bromo-5-methyl,2-bromo-5-methyl pyridine,5-methyl-2-bromopyridine,2-brom-5-methylpyridin,pubchem1185
564216
2-bromo-5-methylpyridine
CC1=CN=C(C=C1)Br
3510-66-5
C6H6BrN
172.02
MFCD00209553
YWNJQQNBJQUKME-UHFFFAOYSA-N
2-bromo-5-picoline,6-bromo-3-picoline,2-bromo-5-methyl-pyridine,pyridine, 2-bromo-5-methyl,2-bromo-5-methyl pyridine,5-methyl-2-bromopyridine,2-brom-5-methylpyridin,pubchem1185
564216
2-bromo-5-methylpyridine
CC1=CN=C(C=C1)Br
Flash Point 103°C
Boiling Point 95°C to 96°C (12.5mmHg)
Color Brown or White to Yellow
Melting Point 40°C to 45°C
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Br(C5H3N)CH3
Formula Weight 172.02
Physical Form Low Melting Crystalline Solid or Crystals
Percent Purity ≥97.5%
Chemical Name or Material 2-Bromo-5-methylpyridine, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

TSCA : TSCA