Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-nitropyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
4487-59-6
C5H3BrN2O2
203.00
MFCD00006222
HUUFTVUBFFESEN-UHFFFAOYSA-N
2-bromo-5-nitro pyridine, 2-bromo-5-nitro-pyridine, 5-nitro-2-bromopyridine, 2-brom-5-nitropyridin, pubchem1183, acmc-1agnm, 2-bromo-5-nitro-pyridin, ksc237s9f, tpc-py095
78240
2-bromo-5-nitropyridine
[O-][N+](=O)C1=CC=C(Br)N=C1
4487-59-6
C5H3BrN2O2
203.00
MFCD00006222
HUUFTVUBFFESEN-UHFFFAOYSA-N
2-bromo-5-nitro pyridine, 2-bromo-5-nitro-pyridine, 5-nitro-2-bromopyridine, 2-brom-5-nitropyridin, pubchem1183, acmc-1agnm, 2-bromo-5-nitro-pyridin, ksc237s9f, tpc-py095
78240
2-bromo-5-nitropyridine
[O-][N+](=O)C1=CC=C(Br)N=C1
Packaging Glass bottle
Color Brown to Yellow
Infrared Spectrum Authentic
Boiling Point 145°C to 147°C (10.0mmHg)
Quantity 1g
Melting Point 137°C to 141°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 203
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-nitropyridine, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-777-2