Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-benzyloxypyridine 97.0+%, TCI America™
SDP

Catalog No. B257625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B257625G 25 g
B25765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B257625G Supplier TCI America Supplier No. B257625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117068-71-0
Molecular Formula C12H10BrNO
Molecular Weight (g/mol) 264.12
MDL Number MFCD00234782
InChI Key WEMPEMXDSUATEK-UHFFFAOYSA-N
PubChem CID 817475
IUPAC Name 2-(benzyloxy)-6-bromopyridine
SMILES BrC1=CC=CC(OCC2=CC=CC=C2)=N1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 264.12
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-6-benzyloxypyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.