Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-fluorobenzaldehyde, 98%

Catalog No. AAH6468914
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 360575-28-6 | C7H4BrFO | 203.01 g/mol

It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 360575-28-6
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD03407341
InChI Key PJNILWKRAKKEQM-UHFFFAOYSA-N
Synonym 6-bromo-2-fluorobenzaldehyde, 2-bromo-6-fluorobenzenecarbaldehyde, 2-bromo-6-fluoro-benzaldehyde, 2-fluoro-6-bromobenzaldehyde, 6-fluoro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-6-fluoro, pubchem1419, acmc-1afpf, ksc497k8t
PubChem CID 22473977
IUPAC Name 2-bromo-6-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Br)C=O)F

Specifications

Melting Point 43°C to 47°C
Density 1.70 g/mL
Boiling Point 100°C to 102°C (8 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2573976
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 203.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-fluorobenzaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.