Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-fluorobenzylamine hydrochloride, 96%

Catalog No. AAH6208606
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 1214330-90-1 | C7H8BrClFN | 240.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214330-90-1
Molecular Formula C7H8BrClFN
Molecular Weight (g/mol) 240.5
InChI Key WEGQVRTUTQKDNY-UHFFFAOYSA-N
Synonym 2-bromo-6-fluorobenzylamine hcl, 2-bromo-6-fluorophenyl methanamine hydrochloride, 2-bromo-6-fluorophenyl methanamine;hydrochloride, 1-2-bromo-6-fluorophenyl methanamine hydrochloride
PubChem CID 118798502
IUPAC Name (2-bromo-6-fluorophenyl)methanamine;hydrochloride
SMILES C1=CC(=C(C(=C1)Br)CN)F.Cl

Specifications

Quantity 5 g
Formula Weight 240.5
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-fluorobenzylamine hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.