Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-nitrophenol, 97%, Thermo Scientific™

Catalog No. ac442680050
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 13073-25-1
Molecular Formula C6H4BrNO3
Molecular Weight (g/mol) 218.01
MDL Number MFCD02683216
InChI Key VEJSIOPQKQXJAT-UHFFFAOYSA-N
Synonym 2-bromo-6-nitrophenol, phenol, 2-bromo-6-nitro, 2-bromo-6-nitro-phenol, 6-bromo-2-nitrophenol, 6-nitro-2-bromophenol, 3-bromo-2-hydroxynitrobenzene, pubchem3114, 2-brom-6-nitrophenol, 2-nitro-6-bromophenol, acmc-209bjb
PubChem CID 13545453
IUPAC Name 2-bromo-6-nitrophenol
SMILES OC1=C(Br)C=CC=C1[N+]([O-])=O

Specifications

Melting Point 67.0°C to 69.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 218.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-nitrophenol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.