Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-m-xylene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
576-22-7
C8H9Br
185.06
MFCD00000075
MYMYVYZLMUEVED-UHFFFAOYSA-N
2,6-dimethylbromobenzene,2-bromo-m-xylene,1-bromo-2,6-dimethylbenzene,benzene, 2-bromo-1,3-dimethyl,2,6-dimethyl bromobenzene,m-xylene, 2-bromo,2,6-dimethylphenyl bromide,2-bromo-meta-xylene,2-bromo-1,3-dimethyl-benzene,pubchem3206
68471
2-bromo-1,3-dimethylbenzene
CC1=C(C(=CC=C1)C)Br
576-22-7
C8H9Br
185.06
MFCD00000075
MYMYVYZLMUEVED-UHFFFAOYSA-N
2,6-dimethylbromobenzene,2-bromo-m-xylene,1-bromo-2,6-dimethylbenzene,benzene, 2-bromo-1,3-dimethyl,2,6-dimethyl bromobenzene,m-xylene, 2-bromo,2,6-dimethylphenyl bromide,2-bromo-meta-xylene,2-bromo-1,3-dimethyl-benzene,pubchem3206
68471
2-bromo-1,3-dimethylbenzene
CC1=C(C(=CC=C1)C)Br
Melting Point -10°C
Flash Point 73°C
Density 1.3800g/mL
Boiling Point 206°C
Specific Gravity 1.38
Refractive Index 1.5540 to 1.556
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2C6H3Br
Beilstein 05, 374
Formula Weight 185.06
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Bromo-m-xylene
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.