Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
o-Toluoyl chloride, 99%
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                5 g
                            
                        
                            
                                100 g
                            
                        
                            
                                500 g
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Glass bottle
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
            Specifications
Specifications
| Color | Pink to Yellow | 
| Density | 1.1800g/mL | 
| Boiling Point | 88°C to 90°C (12.0 mmHg) | 
| Flash Point | 76°C | 
| Infrared Spectrum | Authentic | 
| Assay Percent Range | 99% | 
| Packaging | Glass bottle | 
| Refractive Index | 1.5540 to 1.556 | 
| Linear Formula | CH3C6H4COCl | 
| Quantity | 100 g | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Danger | 
| Hazard Statement | GHS H Statement: Causes severe skin burns and eye damage. Causes serious eye damage. | 
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. | 
| EINECSNumber | 213-273-8 | 
| TSCA | TSCA | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                