Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-p-xylene, 97%

Catalog No. p-77667
Click to view available options
Quantity:
25 g
100 g

CAS: 553-94-6 | C8H9Br | 185.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-94-6
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000074
InChI Key QXISTPDUYKNPLU-UHFFFAOYSA-N
Synonym 2,5-dimethylbromobenzene, 2-bromo-p-xylene, 1-bromo-2,5-dimethylbenzene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, 2-bromo-1,4-dimethyl-benzene, 2,5-xylyl bromide, 2-bromo-1,4-xylene, 2,5-dimethylphenyl bromide, p-xylene, 2-bromo
PubChem CID 11121
IUPAC Name 2-bromo-1,4-dimethylbenzene
SMILES CC1=CC=C(C)C(Br)=C1

Specifications

Melting Point 9°C to 10°C
Color Colorless
CAS Min % 96.0
CAS Max % 100.0
Density 1.3400g/mL
Boiling Point 199°C to 201°C
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5495 to 1.5515
Assay Percent Range 96% min. (GC)
Specific Gravity 1.34
Linear Formula (CH3)2C6H3Br
Beilstein 05, 385
Formula Weight 185.06
Percent Purity 97%
Chemical Name or Material 2-Bromo-p-xylene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 209-055-7
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.