Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoadamantane, 98%

Catalog No. p-7026650
Click to view available options
Quantity:
5 g
25 g

CAS: 7314-85-4 | C10H15Br | 215.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7314-85-4
Molecular Formula C10H15Br
Molecular Weight (g/mol) 215.134
MDL Number MFCD00074745
InChI Key RCXJARRRXOPXBC-UHFFFAOYSA-N
Synonym 2-adamantyl bromide, adamantane, 2-bromo, tricyclo 3.3.1.13,7 decane, 2-bromo, tricyclo 3.3.1.1 3,7 decane, 2-bromo, 1r,3r,5r,7r-2-bromoadamantane, 2-bromo-adamantane, 2-bromanyladamantane, pubchem8752, 2alpha-bromoadamantane, 2-bromotricyclo 3.3.1.13.7 decane
PubChem CID 522482
IUPAC Name 2-bromoadamantane
SMILES C1C2CC3CC1CC(C2)C3Br

Specifications

Melting Point 136°C to 141°C
Quantity 5 g
Beilstein 4655839
Formula Weight 215.14
Percent Purity 98%
Chemical Name or Material 2-Bromoadamantane
EINECSNumber 230-777-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.