Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Bromoallyl)trimethylsilane 90.0+%, TCI America™
SDP

Catalog No. B16955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16955G 5 g
B16951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16955G Supplier TCI America Supplier No. B16955G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81790-10-5
Molecular Formula C6H13BrSi
Molecular Weight (g/mol) 193.159
MDL Number MFCD00043054
InChI Key LUPQCAARZVEFMT-UHFFFAOYSA-N
PubChem CID 4173130
IUPAC Name 2-bromoprop-2-enyl(trimethyl)silane
SMILES C[Si](C)(C)CC(=C)Br

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 193.16
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material (2-Bromoallyl)trimethylsilane
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.