Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzyl bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
3433-80-5
C7H6Br2
249.93
MFCD00000173
LZSYGJNFCREHMD-UHFFFAOYSA-N
2-bromobenzyl bromide,o-bromobenzyl bromide,1-bromo-2-bromomethyl benzene,2-bromobenzylbromide,alpha,2-dibromotoluene,benzene, 1-bromo-2-bromomethyl,alpha,o-dibromotoluene,benzene, bromo bromomethyl,alpha-2-dibromotoluene,a,o-dibromotoluene
76965
1-bromo-2-(bromomethyl)benzene
C1=CC=C(C(=C1)CBr)Br
3433-80-5
C7H6Br2
249.93
MFCD00000173
LZSYGJNFCREHMD-UHFFFAOYSA-N
2-bromobenzyl bromide,o-bromobenzyl bromide,1-bromo-2-bromomethyl benzene,2-bromobenzylbromide,alpha,2-dibromotoluene,benzene, 1-bromo-2-bromomethyl,alpha,o-dibromotoluene,benzene, bromo bromomethyl,alpha-2-dibromotoluene,a,o-dibromotoluene
76965
1-bromo-2-(bromomethyl)benzene
C1=CC=C(C(=C1)CBr)Br
Refractive Index 1.6170 to 1.6190
Color Colorless to Yellow
Packaging Glass bottle
Quantity 25g
Melting Point 29.0°C to 32.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Boiling Point 129.0°C (19.0 mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H4CH2Br
Beilstein 05, 308
Formula Weight 249.93
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material 2-Bromobenzyl bromide, 98%
Show More Show Less

GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 222-334-8

TSCA : TSCA