Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. D151425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D151425G 25 g
D15145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D151425G Supplier TCI America Supplier No. D151425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3433-80-5
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000173
InChI Key LZSYGJNFCREHMD-UHFFFAOYSA-N
Synonym 2-bromobenzyl bromide, o-bromobenzyl bromide, 1-bromo-2-bromomethyl benzene, 2-bromobenzylbromide, alpha,2-dibromotoluene, benzene, 1-bromo-2-bromomethyl, alpha,o-dibromotoluene, benzene, bromo bromomethyl, alpha-2-dibromotoluene, a,o-dibromotoluene
PubChem CID 76965
IUPAC Name 1-bromo-2-(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)Br

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 249.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromobenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.