Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzyl mercaptan, 99%

Catalog No. AAH2624703
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 143888-85-1 | C7H7BrS | 203.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143888-85-1
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.10
MDL Number MFCD04038925
InChI Key UJNSDLRPHRMVGZ-UHFFFAOYSA-N
Synonym 2-bromobenzyl mercaptan, 2-bromobenzylmercaptan, 2-bromophenyl methanethiol, o-bromobenzyl hydrosulfide, 2-bromobenzylthiol, 2-bromophenyl methane-1-thiol, 2-bromobenzyl, 2-bromobenzyl mercap, pubchem6847
PubChem CID 2773673
IUPAC Name (2-bromophenyl)methanethiol
SMILES SCC1=CC=CC=C1Br

Specifications

Density 1.527
Boiling Point 256°C to 257°C
Flash Point >110°C (230°F)
Refractive Index 1.617
Quantity 1 g
UN Number UN2810
Sensitivity Air Sensitive
Formula Weight 203.1
Percent Purity 99%
Chemical Name or Material 2-Bromobenzyl mercaptan
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.