Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B183625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B183625G 25 g
B18365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B183625G Supplier TCI America Supplier No. B183625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5465-63-4
Molecular Formula C7H8BrClN
Molecular Weight (g/mol) 221.50
MDL Number MFCD00012853
InChI Key ZANQNQMPTMEAIP-UHFFFAOYSA-N
Synonym 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2
PubChem CID 2724202
IUPAC Name 1-(2-bromophenyl)methanamine hydrochloridyl
SMILES [Cl].NCC1=CC=CC=C1Br

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 222.51
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromobenzylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.