Alfa Aesar™ 2-Bromoethanol, 97%

Manufacturer: Alfa Aesar™ A1027514

Quantity 25g

 View more versions of this product

Catalog No. AAA1027514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 540-51-2
    Molecular Formula C2H5BrO
    Linear Formula BrCH2CH2OH
    Formula Weight 124.97
    MDL Number MFCD00002827
    UN Number UN2922
    Beilstein 878140
    Merck Index 14,3792
    Synonym Ethylene bromohydrin; 2-Hydroxyethyl bromide
    Chemical Name or Material 2-Bromoethanol

    Specifications

    Formula Weight 124.97
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 540-51-2
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Boiling Point 149° to 150°C
    Linear Formula BrCH2CH2OH
    MDL Number MFCD00002827
    UN Number UN2922
    Beilstein 878140
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    Flash Point >110°C (230°F)
    Molecular Formula C2H5BrO
    Merck Index 14,3792
    Synonym Ethylene bromohydrin; 2-Hydroxyethyl bromide
    RTECS Number KJ8225000
    TSCA TSCA
    Melting Point −80°C
    Quantity 25g
    Chemical Name or Material 2-Bromoethanol
    Assay 97%

    Description

    Applications
    2-Bromoethanol is utilized in selective reduction of nitroarenes. It is used as a solvent and organic synthetic raw material. It finds application in the synthesis of 2-piperidin-1-yl-ethanol by reacting with piperidine. It is used in selective reduction of nitroarenes.

    Solubility
    Miscible with water, organic solvent and petroleum ether.

    Notes
    Light sensitive. Hygroscopic. Store in a cool place. Incompatible with strong acids, strong oxidizing agents, strong reducing agents, acid chlorides and acid anhydrides.

    GHS H Statement
    H300-H310-H330-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit