Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoethyl Ethyl Ether 95.0+%, TCI America™
SDP

Catalog No. B059525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B059525G 25 g
B0595100G 100 g
B0595500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B059525G Supplier TCI America Supplier No. B059525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 592-55-2
Molecular Formula C4H9BrO
Molecular Weight (g/mol) 153.02
MDL Number MFCD00000237
InChI Key MMYKTRPLXXWLBC-UHFFFAOYSA-N
Synonym 2-bromoethyl ethyl ether, 2-ethoxyethyl bromide, 2-bromoethoxyethane, ethane, 1-bromo-2-ethoxy, 2-bromodiethyl ether, 1-ethoxy-2-bromoethane, 1-bromo-2-ethoxy-ethane, ether, 2-bromoethyl ethyl, 2-bromoethylethylether, 1-bromo-3-oxapentane
PubChem CID 61141
IUPAC Name 1-bromo-2-ethoxyethane
SMILES CCOCCBr

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 g
UN Number 2340
Formula Weight 153.02
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromoethyl Ethyl Ether
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.