Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Bromoethyl)phosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. B50131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50131G 1 g
B5013200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50131G Supplier TCI America Supplier No. B50131G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 999-82-6
Molecular Formula C2H6BrO3P
Molecular Weight (g/mol) 188.95
MDL Number MFCD00013957
InChI Key BFKXVERGWXHHIH-UHFFFAOYSA-N
PubChem CID 3613627
IUPAC Name (2-bromoethyl)phosphonic acid
SMILES OP(O)(=O)CCBr

Specifications

Melting Point 88°C
Quantity 1 g
UN Number 3261
Formula Weight 188.94
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Bromoethyl)phosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.