Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoethylamine Hydrobromide, >98.0%, MP Biomedicals™

Catalog No. p-4405057
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 2576-47-8
Molecular Formula C2H7Br2N
Molecular Weight (g/mol) 204.893
InChI Key WJAXXWSZNSFVNG-UHFFFAOYSA-N
Synonym 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
PubChem CID 2774217
IUPAC Name 2-bromoethanamine;hydrobromide
SMILES C(CBr)N.Br

Specifications

Color White
Density 1.581g/cm3
Assay Percent Range >98%
Quantity 25 g
Formula Weight 204.9
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoethylamine Hydrobromide, Quality Control
Recommended Storage Store at Room Temperature (15° to 30°C), dessicate.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.