Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoethylamine hydrobromide, 98+%

Catalog No. p-7023757
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 2576-47-8 | C2H7Br2N | 204.893 g/mol

2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.

Solubility
Soluble in water.

Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2576-47-8
Molecular Formula C2H7Br2N
Molecular Weight (g/mol) 204.893
MDL Number MFCD00012886
InChI Key WJAXXWSZNSFVNG-UHFFFAOYSA-N
Synonym 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
PubChem CID 2774217
IUPAC Name 2-bromoethanamine;hydrobromide
SMILES C(CBr)N.Br

Specifications

Melting Point 172°C to 176°C
Quantity 50 g
Beilstein 3607605
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 204.9
Percent Purity ≥98%
Chemical Name or Material 2-Bromoethylamine hydrobromide
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
EINECSNumber 219-924-2
RTECSNumber KQ8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.