Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromoisophthalaldehyde, 97%

Click to view available options
Quantity:
250mg
1g
5g
79839-49-9
C8H5BrO2
213.03
MFCD08461034
RZUSSKMZLHKMHU-UHFFFAOYSA-N
2-bromoisophthalaldehyde,2-bromobenzene-1,3-dialdehyde,2-bromo-1,3-diformylbenzene,2,6-diformylbromobenzene,2-bromo-benzene-1,3-dicarbaldehyde,2-bromobenzene-1,3-dicarboxaldehyde,1,3-diformyl-2-bromobenzene,1,3-benzenedicarboxaldehyde, 2-bromo,acmc-209pia,ksc495c0d
11435812
2-bromobenzene-1,3-dicarbaldehyde
C1=CC(=C(C(=C1)C=O)Br)C=O
79839-49-9
C8H5BrO2
213.03
MFCD08461034
RZUSSKMZLHKMHU-UHFFFAOYSA-N
2-bromoisophthalaldehyde,2-bromobenzene-1,3-dialdehyde,2-bromo-1,3-diformylbenzene,2,6-diformylbromobenzene,2-bromo-benzene-1,3-dicarbaldehyde,2-bromobenzene-1,3-dicarboxaldehyde,1,3-diformyl-2-bromobenzene,1,3-benzenedicarboxaldehyde, 2-bromo,acmc-209pia,ksc495c0d
11435812
2-bromobenzene-1,3-dicarbaldehyde
C1=CC(=C(C(=C1)C=O)Br)C=O
Applications
Used in the preparation of paclitaxel-mimicking alkaloids with anticancer activity.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 5g
Melting Point 139°C to 144°C
Beilstein 4740331
Formula Weight 213.03
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Bromoisophthalaldehyde

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger