Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester, 95%, Thermo Scientific Chemicals

Catalog No. AAH5330903
Click to view available options
Quantity:
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 441011-76-3
Molecular Formula C12H16BBrO2
MDL Number MFCD04039022
Synonym 2-(Bromomethyl)phenylboronic acid neopentyl glycol ester; 2-(Bromomethyl)phenylboronic acid mono(2,2-dimethylpropyl) ester

Specifications

Quantity 1 g
Formula Weight 282.97
Percent Purity 95%
Chemical Name or Material 2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.