Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Bromomethyl)benzeneboronic acid pinacol ester, 98%

Manufacturer:  Alfa Aesar™ H5321703

Quantity 1g

 View more versions of this product

Catalog No. AAH5321703


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

377780-72-8
297
2-(Bromomethyl)phenylboronic acid pinacol ester; 2-[2-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C13H18BBrO2
MFCD01632210
2-(Bromomethyl)benzeneboronic acid pinacol ester

Specifications

377780-72-8
297
78° to 80°C
1g
98%
C13H18BBrO2
MFCD01632210
2-(Bromomethyl)phenylboronic acid pinacol ester; 2-[2-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(Bromomethyl)benzeneboronic acid pinacol ester
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit