Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Bromomethyl)benzeneboronic acid pinacol ester, 98%

Catalog No. p-7035500
Click to view available options
Quantity:
250 mg
1 g

Catalog No. AAH53217MD

Supplier: Thermo Scientific Chemicals H53217MD

Only null left
Add to Cart

CAS: 377780-72-8 | C13H18BBrO2 | 296.999 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 377780-72-8
Molecular Formula C13H18BBrO2
Molecular Weight (g/mol) 296.999
MDL Number MFCD01632210
InChI Key ROIXSNLOYHDYBP-UHFFFAOYSA-N
Synonym 2-2-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromomethylphenylboronic acid pinacol ester, 2-bromomethyl benzeneboronic acid, pinacol ester, 2-bromomethylphenylboronic acid, pinacol ester, 2-bromomethyl benzeneboronic acid pinacol ester, 2-bromomethylphenyl boronic acid, pinacol ester, amtb373, 2-bromomethyl phenylboronic pinacol ester, 2-bromoethylphenylboronic acid pinacol ester
PubChem CID 3834834
IUPAC Name 2-[2-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2CBr

Specifications

Melting Point 78°C to 80°C
Quantity 250 mg
Formula Weight 297
Percent Purity 98%
Chemical Name or Material 2-(Bromomethyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.