Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Bromomethyl)oxetane, 96%

Click to view available options
Quantity:
250mg
1g
939759-23-6
C4H7BrO
151.003
MFCD08544405
OMXAGUVERXNCSZ-UHFFFAOYSA-N
2-bromomethyl oxetane,2-bromomethyl-oxetane,oxetane, 2-bromomethyl,2-bromomethyl oxetane,cyclobutane, bromomethyl,2-bromomethyl oxetane oxetane, 2-bromomethyl
16244494
2-(bromomethyl)oxetane
C1COC1CBr
939759-23-6
C4H7BrO
151.003
MFCD08544405
OMXAGUVERXNCSZ-UHFFFAOYSA-N
2-bromomethyl oxetane,2-bromomethyl-oxetane,oxetane, 2-bromomethyl,2-bromomethyl oxetane,cyclobutane, bromomethyl,2-bromomethyl oxetane oxetane, 2-bromomethyl
16244494
2-(bromomethyl)oxetane
C1COC1CBr
Color Colorless to Yellow
Quantity 250mg
Sensitivity Moisture Sensitive
Formula Weight 151
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 2-(Bromomethyl)oxetane