Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromonaphthalene, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
580-13-2
C10H7Br
207.07
APSMUYYLXZULMS-UHFFFAOYSA-N
naphthalene, 2-bromo,2-naphthyl bromide,2-bromo-naphthalene,beta-bromonaphthalene,beta-naphthyl bromide,.beta.-bromonaphthalene,unii-syu33i753n,naphthalene,2-bromo,.beta.-naphthyl bromide,pbn-2
11372
2-bromonaphthalene
C1=CC=C2C=C(C=CC2=C1)Br
580-13-2
C10H7Br
207.07
APSMUYYLXZULMS-UHFFFAOYSA-N
naphthalene, 2-bromo,2-naphthyl bromide,2-bromo-naphthalene,beta-bromonaphthalene,beta-naphthyl bromide,.beta.-bromonaphthalene,unii-syu33i753n,naphthalene,2-bromo,.beta.-naphthyl bromide,pbn-2
11372
2-bromonaphthalene
C1=CC=C2C=C(C=CC2=C1)Br
Color Brown-Orange to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Boiling Point 281.0°C to 282.0°C
Quantity 5g
Packaging Glass bottle
Melting Point 53.0°C to 57.0°C
Assay Percent Range 98.5% min. (GC)
Linear Formula C10H7Br
Beilstein 05,548
Merck Index 15,1437
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in 8 parts alcohol,ether,chloroform and,benzene
Formula Weight 207.07
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Bromonaphthalene
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 209-452-5