Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyl acetate, 99%, pure

Catalog No. AC158290025
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 109-60-4 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009372
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
Synonym n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester
PubChem CID 7997
ChEBI CHEBI:40116
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -95.0°C
Density 0.8800g/mL
Boiling Point 102.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3830 to 1.3850
Linear Formula CH3CO2CH2CH2CH3
Quantity 2.5 L
UN Number 1276
Beilstein 02, 129
Specific Gravity 0.88
Merck Index 15, 7954
Solubility Information Solubility in water: 2g/100mL (20°C). Other solubilities: miscible with alcohols,ketones,esters and,hydrocarbons
Viscosity 0.58 mPa.s (20°C)
Formula Weight 102.13
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Propyl acetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
EINECSNumber 203-686-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.