Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromophenyl isocyanate, 97%, Thermo Scientific™

Catalog No. AC305530010
Click to view available options
:
1g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1592-00-3
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H4BrNO
Refractive Index 1.5820 to 1.5860 (20°C, 589nm)
Linear Formula BrC6H4NCO
Quantity 1g
MDL Number MFCD00001995
Synonym 2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate
InChI Key GOOVAYJIVMBWPP-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)N=C=O)Br
IUPAC Name 1-bromo-2-isocyanatobenzene
Molecular Weight (g/mol) 198.019
PubChem CID 137097
Formula Weight 198.02
Percent Purity ≥96.0%
Chemical Name or Material 2-Bromophenyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is co
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.