Learn More
(2-Bromophenylethynyl)trimethylsilane, 98%
Description
(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Boiling Point | 60°C to 80°C (2mmHg) |
| Quantity | 25 g |
| Flash Point | >113°C (235°F) |
| Solubility Information | Slightly soluble in water. |
| IUPAC Name | 2-(2-bromophenyl)ethynyl-trimethylsilane |
| Formula Weight | 253.21 |
| Percent Purity | 98% |
| Refractive Index | 1.554 |
| Density | 1.19 |
| Chemical Name or Material | (2-Bromophenylethynyl)trimethylsilane |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.