Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromopropane, 99%

Manufacturer:  Alfa Aesar A12944AK

Quantity 250mL

 View more versions of this product

Catalog No. AAA12944AK


  Check Availability
Add to cart

Specifications

Description

Applications
2-Bromopropane serves as an alkylating agent in organic synthesis. It is also used as an intermediate to form alkylated amines and alkylated metallic compounds. Further, it acts as a solvent for industrial cleaning, degreasing, metal processing and finishing, electronics, aerospace and aviation, aerosols, textiles, adhesives and inks. In addition, it is used for introducing the isopropyl functional group in organic synthesis.

Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water and acetone.

Notes
Avoid all possible sources of ignition.Incompatible with strong oxidizing agents and strong bases.
Description & Specifications

Chemical Identifiers

75-26-3
CH3CHBrCH3
MFCD00000147
741852
2-Bromopropane
CH3CHBrCH3
123
UN2344
14,5210

Specifications

75-26-3
250mL
99%
CH3CHBrCH3
MFCD00000147
741852
2-Bromopropane
1.310
Liquid
CH3CHBrCH3
123
UN2344
14,5210
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H360-H373

GHS P Statement
P210-P260-P201-P280-P281-P240-P241-P303+P361+P353-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BROMOPROPANES

RTECSNumber : TX4111000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit