Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromopropionyl bromide, 97%

Catalog No. p-7026487
Click to view available options
Quantity:
100 g
500 g

CAS: 563-76-8 | C3H4Br2O | 215.872 g/mol

2-Bromopropionyl bromide is used in the preparation of cellulose macroinitiator through direct acylation of cellulose in ionic liquid 1-allyl-3-methylimidazolium chloride. It is also used to prepare 3-(2-bromopropionyl)-2-oxazolidone derivatives and bromoester terminated poly(3-hexylthiophene). Further, it is used as a macroinitiator for atom transfer radical polymerization of acrylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopropionyl bromide is used in the preparation of cellulose macroinitiator through direct acylation of cellulose in ionic liquid 1-allyl-3-methylimidazolium chloride. It is also used to prepare 3-(2-bromopropionyl)-2-oxazolidone derivatives and bromoester terminated poly(3-hexylthiophene). Further, it is used as a macroinitiator for atom transfer radical polymerization of acrylates.

Solubility
Miscible with trichloromethane, diethyl ether, benzene and acetone.

Notes
Incompatible with water, alcohols, oxidizing agents and strong alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-76-8
Molecular Formula C3H4Br2O
Molecular Weight (g/mol) 215.872
MDL Number MFCD00000142
InChI Key ILLHORFDXDLILE-UHFFFAOYSA-N
Synonym 2-bromopropionyl bromide, propanoyl bromide, 2-bromo, 2-bromopropionylbromide, 2-bromo propionyl bromide, 2-bromopropionic acid bromide, .alpha.-bromopropionyl bromide, 2-bromopropanyl bromide, a-bromopropionyl bromide, 2-bromo-propionyl bromide, alpha-bromopropionylbromide
PubChem CID 79047
IUPAC Name 2-bromopropanoyl bromide
SMILES CC(C(=O)Br)Br

Specifications

Density 2.061
Boiling Point 48°C to 50°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.521
Quantity 100 g
UN Number UN3265
Beilstein 1071331
Sensitivity Moisture sensitive
Solubility Information Miscible with trichloromethane,diethyl ether,benzene and acetone.
Formula Weight 215.88
Percent Purity 97%
Chemical Name or Material 2-Bromopropionyl bromide
Show More Show Less
Hazard Category H302-H314-H317-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 209-261-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.