Learn More
2-Bromopyridine, 99%
Description
2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.
Solubility
Slightly miscible with water.
Notes
Keep away from heat, sparks, and flame. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and acid chlorides.

Specifications
Specifications
Density | 1.625 |
Boiling Point | 192°C to 194°C |
Flash Point | 97°C (207°F) |
Odor | Characteristic |
Refractive Index | 1.572 |
Quantity | 250 g |
UN Number | UN2810 |
Beilstein | 105789 |
Solubility Information | Slightly miscible with water. |
Formula Weight | 158 |
Show More |
Safety and Handling
Hazard Category | H226-H301+H331-H310-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H310-H315-H319-H335 Toxic if swallowed. Fatal in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P332+P313-P362-P374-P380-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
EINECSNumber | 203-641-6 |
RTECSNumber | US3850000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.