Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Bromothiophene, 98%
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -10.0°C |
Color | Brown to Colorless |
Density | 1.6800g/mL |
Boiling Point | 149.0°C to 151.0°C |
Flash Point | 52°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass Bottle |
Refractive Index | 1.5850 to 1.5870 |
Quantity | 1 L |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. Toxic if swallowed. Fatal in contact with skin. Fatal if inhaled. Flammable liquid and vapour. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses |
EINECSNumber | 213-699-4 |
Recommended Storage | Product darkens on exposure to light |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction