Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromothiophenol, 97%

Catalog No. p-3777738
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle

CAS: 6320-02-1 | C6H5BrS | 189.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6320-02-1
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00004827
InChI Key YUQUNWNSQDULTI-UHFFFAOYSA-N
Synonym 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol
PubChem CID 80599
SMILES SC1=CC=CC=C1Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6040g/mL
Boiling Point 114°C to 116°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6345 to 1.6365
Linear Formula BrC6H4SH
Quantity 1 g
Beilstein 06, II, 300
Specific Gravity 1.604
Solubility Information Solubility in water: immiscible
IUPAC Name 2-chlorobenzenethiol
Formula Weight 189.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromothiophenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Toxic if inhaled.
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 228-665-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.